The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272090.1 | NP_001259019.1 | 1 - 2 | 1232 | 80 |
NM_001272092.1 | NP_001259021.1 | 1 - 2 | 1232 | 80 | |
NM_027912.1 | NP_082188.1 | 1 - 2 | 1232 | 80 | |
GenBank mRNA | AK005069.1 | - | 1 - 2 | 1233 | 80 |
AK154513.1 | - | 1 - 2 | 1215 | 80 | |
BC119320.1 | - | 1 - 2 | 1210 | 80 |