Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr55
Entrez Gene ID
67936
Gene Name
WD repeat domain 55
Gene Aliases
2410080P20Rik, C80692
Chromosome Location
Chr.18: 36759739 - 36763708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026464.2 NP_080740.2 4 - 5 613 92
GenBank mRNA AK018469.1 - 4 - 5 594 92
AK075958.1 - 4 - 5 613 92
AK146093.1 - 4 - 5 632 92
BC011484.1 - 2 - 3 279 92
BC064750.1 - 4 - 5 572 92
BC131932.1 - 4 - 5 607 92
BC131934.1 - 4 - 5 607 92

View Full Product Details