Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cul1
Entrez Gene ID
26965
Gene Name
cullin 1
Gene Aliases
-
Chromosome Location
Chr.6: 47452273 - 47526139 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012042.3 NP_036172.1 5 - 6 856 141
GenBank mRNA AF136441.1 - 4 - 5 711 141
AF176910.1 - 5 - 6 745 141
AK147004.1 - 5 - 6 838 141
AK164274.1 - 5 - 6 823 141
BC004836.1 - 4 - 5 665 141
BC029260.1 - 5 - 6 856 141

View Full Product Details