Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca14
Entrez Gene ID
67928
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 14
Gene Aliases
1700110B15Rik, 4930539G24Rik
Chromosome Location
Chr.7: 120203895 - 120325352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026458.4 NP_080734.3 21 - 22 3558 95
GenBank mRNA AY243470.1 - 21 - 22 3558 95

View Full Product Details