The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026458.4 | NP_080734.3 | 9 - 10 | 1423 | 92 |
GenBank mRNA | AK029665.1 | - | 10 - 11 | 1535 | 92 |
AY243470.1 | - | 9 - 10 | 1423 | 92 |