The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026617.3 | NP_080893.1 | 6 - 7 | 526 | 69 |
GenBank mRNA | AK006207.1 | - | 6 - 7 | 527 | 69 |
AK153442.1 | - | 5 - 6 | 454 | 69 | |
AK184146.1 | - | 4 - 5 | 291 | 69 |