Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan17
Entrez Gene ID
74257
Gene Name
tetraspanin 17
Gene Aliases
2210021G21Rik, AI047581, Fbxo23, Tm4sf17
Chromosome Location
Chr.13: 54789352 - 54796896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CD740983.1 - 1 - 2 61

View Full Product Details