The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146205.2 | NP_666317.2 | 1 - 2 | 1232 | 63 |
GenBank mRNA | AK030000.1 | - | 1 - 2 | 89 | 63 |
AK131111.1 | - | 1 - 2 | 1232 | 63 | |
BC032200.1 | - | 1 - 2 | 577 | 63 | |
BC089363.1 | - | 1 - 2 | 473 | 63 |