The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130163.1 | NP_001123635.1 | 5 - 6 | 566 | 135 |
NM_001130165.1 | NP_001123637.1 | 5 - 6 | 661 | 135 | |
NM_001130166.1 | NP_001123638.1 | 7 - 8 | 925 | 135 | |
NM_130885.2 | NP_570955.1 | 5 - 6 | 566 | 135 | |
GenBank mRNA | AF324899.1 | - | 5 - 6 | 565 | 135 |
AF333985.1 | - | 5 - 6 | 566 | 135 | |
AK040881.1 | - | 5 - 6 | 589 | 135 | |
AK052714.1 | - | 5 - 6 | 731 | 135 | |
AK158703.1 | - | 7 - 8 | 907 | 135 | |
BC089183.1 | - | 5 - 6 | 656 | 135 | |
BC098491.1 | - | 7 - 8 | 908 | 135 |