The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010826.5 | NP_034956.2 | 12 - 13 | 1871 | 75 |
NM_194464.3 | NP_919446.2 | 13 - 14 | 2114 | 75 | |
GenBank mRNA | AK141821.1 | - | 8 - 9 | 1153 | 75 |
AK157169.1 | - | 13 - 14 | 1724 | 75 | |
U63407.1 | - | 13 - 14 | 2117 | 75 | |
U63408.1 | - | 12 - 13 | 1866 | 75 |