Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfc4
Entrez Gene ID
106344
Gene Name
replication factor C (activator 1) 4
Gene Aliases
A1, AI894123, AU040575, RFC37
Chromosome Location
Chr.16: 23113948 - 23127730 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145480.1 NP_663455.1 10 - 11 1059 141
GenBank mRNA AK146954.1 - 10 - 11 1047 141
BC003335.1 - 10 - 11 1059 141

View Full Product Details