The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190911.1 | NP_001177840.1 | 5 - 6 | 966 | 79 |
NM_001190912.1 | NP_001177841.1 | 4 - 5 | 888 | 79 | |
NM_001190913.1 | NP_001177842.1 | 5 - 6 | 966 | 79 | |
NM_001190914.1 | NP_001177843.1 | 6 - 7 | 1067 | 79 | |
NM_026324.3 | NP_080600.1 | 5 - 6 | 966 | 79 | |
GenBank mRNA | AK034142.1 | - | 5 - 6 | 903 | 79 |
AK038385.1 | - | 5 - 6 | 717 | 79 | |
AK045373.1 | - | 5 - 6 | 871 | 79 | |
AK079332.1 | - | 6 - 7 | 1005 | 79 | |
AK171866.1 | - | 2 - 3 | 317 | 79 | |
AY169782.1 | - | 5 - 6 | 966 | 79 | |
BC063072.1 | - | 4 - 5 | 683 | 79 |