The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190911.1 | NP_001177840.1 | 6 - 7 | 1121 | 66 |
NM_001190912.1 | NP_001177841.1 | 5 - 6 | 1043 | 66 | |
NM_001190913.1 | NP_001177842.1 | 6 - 7 | 1121 | 66 | |
NM_001190914.1 | NP_001177843.1 | 7 - 8 | 1222 | 66 | |
NM_026324.3 | NP_080600.1 | 6 - 7 | 1121 | 66 | |
GenBank mRNA | AK034142.1 | - | 6 - 7 | 1058 | 66 |
AK038385.1 | - | 6 - 7 | 872 | 66 | |
AK045373.1 | - | 6 - 7 | 1026 | 66 | |
AK079332.1 | - | 7 - 8 | 1160 | 66 | |
AK171866.1 | - | 3 - 4 | 472 | 66 | |
AY169782.1 | - | 6 - 7 | 1121 | 66 | |
BC063072.1 | - | 5 - 6 | 838 | 66 |