Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Angptl6
Entrez Gene ID
70726
Gene Name
angiopoietin-like 6
Gene Aliases
6330404E11Rik, AGF, ARP3, Arp5
Chromosome Location
Chr.9: 20873808 - 20879816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145154.2 NP_660136.1 1 - 2 110 97

View Full Product Details