The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134383.1 | NP_001127855.1 | 2 - 3 | 610 | 86 |
NM_001134384.1 | NP_001127856.1 | 2 - 3 | 644 | 86 | |
GenBank mRNA | AK157963.1 | - | 2 - 3 | 663 | 86 |