Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xpc
Entrez Gene ID
22591
Gene Name
xeroderma pigmentosum, complementation group C
Gene Aliases
-
Chromosome Location
Chr.6: 91489305 - 91515888 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009531.2 NP_033557.2 6 - 7 854 63
GenBank mRNA AB071144.1 - 6 - 7 791 63
AK004713.1 - 6 - 7 794 63
AK028595.1 - 6 - 7 792 63
AK166981.1 - 6 - 7 789 63
AK170975.1 - 6 - 7 786 63
BC052725.1 - 6 - 7 816 63
U27398.1 - 6 - 7 762 63
U40005.1 - 5 - 6 593 63

View Full Product Details