The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256104.1 | NP_001243033.1 | 1 - 2 | 156 | 79 |
GenBank mRNA | BC023050.1 | - | 1 - 2 | 101 | 79 |
CN723057.1 | - | 1 - 2 | 38 | 79 |