Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcea3
Entrez Gene ID
21401
Gene Name
transcription elongation factor A (SII), 3
Gene Aliases
S-II, SII-K1
Chromosome Location
Chr.4: 136247936 - 136274903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011542.2 NP_035672.1 4 - 5 470 82
GenBank mRNA AJ223472.1 - 4 - 5 472 82
AK002319.1 - 4 - 5 454 82
AK007437.1 - 4 - 5 476 82
BC010807.1 - 4 - 5 463 82
D83387.1 - 4 - 5 412 82

View Full Product Details