Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd4
Entrez Gene ID
245469
Gene Name
PDZ domain containing 4
Gene Aliases
B230341P03, D130075J17Rik, Kiaa1444-hp, LU1, PDZRN4L, Pdzk4, Pdzrnk4l, Xlu
Chromosome Location
Chr.X: 73793357 - 73824969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029868.2 NP_001025039.1 3 - 4 1031 71
GenBank mRNA BC056462.1 - 3 - 4 377 71

View Full Product Details