Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dctd
Entrez Gene ID
320685
Gene Name
dCMP deaminase
Gene Aliases
6030466N05Rik
Chromosome Location
Chr.8: 48099092 - 48141667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178788.4 NP_848903.2 1 - 2 290 91
NR_027759.1 1 - 2 290 91
GenBank mRNA AK031645.1 - 1 - 2 255 91
AK077663.1 - 1 - 2 252 91
AK171894.1 - 1 - 2 87 91
BB621273.1 - 1 - 2 255 91
BC031719.1 - 1 - 2 74 91

View Full Product Details