Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ext1
Entrez Gene ID
14042
Gene Name
exostoses (multiple) 1
Gene Aliases
AA409028
Chromosome Location
Chr.15: 53068261 - 53346183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010162.2 NP_034292.2 3 - 4 1980 100
GenBank mRNA AK132327.1 - 3 - 4 1957 100
BC004741.1 - 3 - 4 1344 100
U78539.1 - 3 - 4 1484 100
X96639.1 - 3 - 4 1754 100

View Full Product Details