The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008391.4 | NP_032417.3 | 8 - 9 | 934 | 61 |
GenBank mRNA | AK087926.1 | - | 7 - 8 | 802 | 61 |
AK132099.1 | - | 8 - 9 | 947 | 61 | |
AK155025.1 | - | 8 - 9 | 919 | 61 | |
BC006577.1 | - | 8 - 9 | 927 | 61 | |
BC110666.1 | - | 8 - 9 | 906 | 61 | |
J03168.1 | - | 8 - 9 | 889 | 61 |