The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199677.1 | NP_001186606.1 | 1 - 2 | 208 | 67 |
GenBank mRNA | AK158287.1 | - | 1 - 2 | 138 | 67 |
AY081953.2 | - | 1 - 2 | 208 | 67 | |
BC044789.1 | - | 1 - 2 | 111 | 67 |