The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034898.2 | NP_001030070.1 | 3 - 4 | 437 | 76 |
GenBank mRNA | AK143446.1 | - | 3 - 4 | 438 | 76 |
BC147202.1 | - | 3 - 4 | 434 | 76 | |
BC172006.1 | - | 3 - 4 | 498 | 76 | |
BC172010.1 | - | 3 - 4 | 512 | 76 |