The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142642.1 | NP_001136114.1 | 5 - 6 | 996 | 85 |
GenBank mRNA | BC059932.1 | - | 5 - 6 | 979 | 85 |