Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2c44
Entrez Gene ID
226143
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 44
Gene Aliases
AW107714, BC034834, b2b386Clo
Chromosome Location
Chr.19: 44005022 - 44029247 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001446.3 NP_001001446.3 3 - 4 553 92
NM_001167905.1 NP_001161377.1 3 - 4 553 92
GenBank mRNA AK050436.1 - 3 - 4 515 92
AK149451.1 - 3 - 4 515 92
BC022141.1 - 3 - 4 502 92
BC024068.1 - 3 - 4 473 92
BC025819.1 - 3 - 4 505 92
BC026766.1 - 3 - 4 505 92
BC034834.1 - 3 - 4 498 92

View Full Product Details