The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009838.2 | NP_033968.1 | - | 286 | 83 |
GenBank mRNA | AK077916.1 | - | - | 239 | 83 |
AK134428.1 | - | - | 239 | 83 | |
AK145522.1 | - | - | 245 | 83 | |
AK147130.1 | - | - | 283 | 83 | |
AK149757.1 | - | - | 645 | 83 | |
AK150977.1 | - | - | 240 | 83 | |
AK159354.1 | - | - | 240 | 83 | |
AK159785.1 | - | - | 260 | 83 | |
AK159975.1 | - | - | 286 | 83 | |
AK160067.1 | - | - | 240 | 83 | |
AK164312.1 | - | - | 239 | 83 | |
AK165612.1 | - | - | 242 | 83 | |
AK166749.1 | - | - | 239 | 83 | |
AK166795.1 | - | - | 269 | 83 | |
AK166870.1 | - | - | 237 | 83 | |
AK166887.1 | - | - | 238 | 83 | |
AK166929.1 | - | - | 238 | 83 | |
AK167007.1 | - | - | 268 | 83 | |
AK167076.1 | - | - | 237 | 83 | |
AK167082.1 | - | - | 235 | 83 | |
AK167164.1 | - | - | 245 | 83 | |
AK167180.1 | - | - | 272 | 83 | |
AK167307.1 | - | - | 240 | 83 | |
AK167669.1 | - | - | 269 | 83 | |
AK167851.1 | - | - | 240 | 83 | |
AK168065.1 | - | - | 235 | 83 | |
AK168940.1 | - | - | 269 | 83 | |
AK169033.1 | - | - | 219 | 83 | |
AK216762.1 | - | - | 127 | 83 | |
BC055043.1 | - | - | 248 | 83 | |
BC069917.1 | - | - | 241 | 83 | |
BC094352.1 | - | - | 212 | 83 | |
Z31557.1 | - | - | 257 | 83 |