The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253877.1 | NP_001240806.1 | 6 - 7 | 1129 | 73 |
NM_001253878.1 | NP_001240807.1 | 6 - 7 | 848 | 73 | |
NM_001253879.1 | NP_001240808.1 | 5 - 6 | 771 | 73 | |
NM_001253880.1 | NP_001240809.1 | 3 - 4 | 486 | 73 | |
NM_013827.3 | NP_038855.2 | 5 - 6 | 767 | 73 | |
NR_045608.1 | 7 - 8 | 835 | 73 | ||
GenBank mRNA | AB221622.1 | - | 2 - 3 | 174 | 73 |
AK030752.1 | - | 5 - 6 | 717 | 73 | |
AK031409.1 | - | 7 - 8 | 836 | 73 | |
AK032798.1 | - | 6 - 7 | 1090 | 73 | |
AK032819.1 | - | 6 - 7 | 1091 | 73 | |
AK137539.1 | - | 7 - 8 | 1076 | 73 | |
AK139685.1 | - | 6 - 7 | 849 | 73 | |
BC024889.1 | - | 5 - 6 | 739 | 73 | |
BC029076.1 | - | 5 - 6 | 678 | 73 | |
BC050826.1 | - | 5 - 6 | 721 | 73 | |
BC057163.1 | - | 5 - 6 | 564 | 73 | |
BC071223.1 | - | 3 - 4 | 239 | 73 | |
BC092237.1 | - | 5 - 6 | 699 | 73 | |
BC100340.1 | - | 3 - 4 | 417 | 73 | |
S78454.1 | - | 2 - 3 | 174 | 73 |