Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam49b
Entrez Gene ID
223601
Gene Name
family with sequence similarity 49, member B
Gene Aliases
0910001A06Rik, AW122079
Chromosome Location
Chr.15: 63929094 - 64060452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144846.5 NP_659095.1 9 - 10 1014 86
GenBank mRNA AK028062.1 - 9 - 10 1015 86
AK075613.1 - 10 - 11 959 86
BC011343.1 - 9 - 10 832 86

View Full Product Details