The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008183.3 | NP_032209.1 | 2 - 3 | 154 | 91 |
GenBank mRNA | BC019426.1 | - | 2 - 3 | 124 | 91 |
BC037068.1 | - | 2 - 3 | 178 | 91 | |
J04696.1 | - | 2 - 3 | 115 | 91 |