Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cux2
Entrez Gene ID
13048
Gene Name
cut-like homeobox 2
Gene Aliases
1700051K22Rik, Cutl2, Cux-2, ENSMUSG00000072641
Chromosome Location
Chr.5: 121857962 - 122050562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007804.2 NP_031830.2 19 - 20 3215 163
GenBank mRNA BC065113.1 - 19 - 20 3215 163
U45665.1 - 19 - 20 3036 163

View Full Product Details