The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290452.1 | NP_001277381.1 | 1 - 2 | 284 | 74 |
NM_001290453.1 | NP_001277382.1 | 1 - 2 | 284 | 74 | |
NM_145819.2 | NP_665818.2 | 1 - 2 | 284 | 74 | |
GenBank mRNA | AB088478.1 | - | 1 - 2 | 185 | 74 |
AK145014.1 | - | 1 - 2 | 165 | 74 | |
BC060649.1 | - | 1 - 2 | 244 | 74 |