The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290452.1 | NP_001277381.1 | 3 - 4 | 790 | 60 |
NM_001290453.1 | NP_001277382.1 | 4 - 5 | 974 | 60 | |
NM_145819.2 | NP_665818.2 | 4 - 5 | 974 | 60 | |
GenBank mRNA | AB007407.1 | - | 3 - 4 | 1193 | 60 |
AB088478.1 | - | 4 - 5 | 875 | 60 | |
AK030410.1 | - | 4 - 5 | 975 | 60 | |
AK158837.1 | - | 2 - 3 | 2226 | 60 | |
BC060649.1 | - | 4 - 5 | 934 | 60 | |
BC129904.1 | - | 3 - 4 | 540 | 60 | |
BC129905.1 | - | 4 - 5 | 724 | 60 |