Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl6
Entrez Gene ID
30840
Gene Name
F-box and leucine-rich repeat protein 6
Gene Aliases
AU021795, Fbl6
Chromosome Location
Chr.15: 76535723 - 76538746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013909.2 NP_038937.2 3 - 4 646 65
GenBank mRNA AB041571.1 - 2 - 3 884 65
AF176522.1 - 3 - 4 634 65
AK040458.1 - 1 - 2 1026 65
AK052585.1 - 2 - 3 885 65
AK183822.1 - 2 - 3 136 65
BC011485.1 - 1 - 2 1019 65
BC026818.1 - 1 - 2 1013 65
BC032263.1 - 6 - 7 65
BC044784.2 - 2 - 3 846 65

View Full Product Details