Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dspp
- Entrez Gene ID
-
666279
- Gene Name
- dentin sialophosphoprotein
- Gene Aliases
-
Dmp2, Dmp3, Dpp, Dsp
- Chromosome Location
-
Chr.5: 104170712 - 104180127 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U67916.1 |
- |
1 - 2 |
65 |
125 |
View Full Product Details