Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Evi5
Entrez Gene ID
14020
Gene Name
ecotropic viral integration site 5
Gene Aliases
NB4S
Chromosome Location
Chr.5: 107744795 - 107875107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007964.2 NP_031990.2 6 - 7 964 109
GenBank mRNA AK134294.1 - 6 - 7 1165 109
AK164899.1 - 3 - 4 289 109
AK165332.1 - 7 - 8 957 109
AK165885.1 - 6 - 7 917 109
BC030056.1 - 7 - 8 971 109
U53586.1 - 6 - 7 964 109

View Full Product Details