Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Golga3
Entrez Gene ID
269682
Gene Name
golgi autoantigen, golgin subfamily a, 3
Gene Aliases
5330413L04, 5430416E01Rik, AI449376, AW490576, G1-499-14, Mea-2, Mea2, repro27
Chromosome Location
Chr.5: 110176659 - 110226470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008146.3 NP_032172.3 15 - 16 3393 73
GenBank mRNA BC043452.1 - 7 - 8 1255 73
BC053002.1 - 15 - 16 3393 73
D78270.2 - 15 - 16 3199 73

View Full Product Details