Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam1b
Entrez Gene ID
280667
Gene Name
a disintegrin and metallopeptidase domain 1b
Gene Aliases
Adam1, Ftna
Chromosome Location
Chr.5: 121500097 - 121502980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172125.2 NP_742123.2 1 - 1 2078 99
GenBank mRNA AK016558.1 - 1 - 1 2533 99
AK161264.1 - 1 - 1 1839 99
BC140979.1 - 1 - 1 2170 99

View Full Product Details