The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093754.2 | NP_001087223.1 | 4 - 5 | 644 | 83 |
NM_001278941.1 | NP_001265870.1 | 5 - 6 | 723 | 83 | |
NM_028110.2 | NP_082386.2 | 4 - 5 | 642 | 83 | |
GenBank mRNA | AK008546.1 | - | 4 - 5 | 448 | 83 |
AK050033.1 | - | 5 - 6 | 724 | 83 | |
AK157279.1 | - | 4 - 5 | 645 | 83 | |
BC008266.1 | - | 4 - 5 | 632 | 83 | |
BC043727.1 | - | 4 - 5 | 430 | 83 |