The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276684.1 | NP_001263613.1 | 2 - 3 | 2137 | 145 |
NM_018790.3 | NP_061260.1 | 2 - 3 | 2140 | 145 | |
GenBank mRNA | AF162777.1 | - | 2 - 3 | 2131 | 145 |
BC023127.1 | - | 2 - 3 | 2126 | 145 |