The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028379.5 | NP_082655.1 | 7 - 8 | 998 | 76 |
GenBank mRNA | AK007572.1 | - | 5 - 6 | 983 | 76 |
AK013609.1 | - | 7 - 8 | 985 | 76 | |
AK156167.1 | - | 7 - 8 | 995 | 76 | |
BC019523.1 | - | 7 - 8 | 980 | 76 | |
BC022770.1 | - | 1 - 2 | 72 | 76 | |
BE853198.1 | - | 1 - 2 | 76 |