The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133712.2 | NP_598473.1 | 5 - 6 | 812 | 72 |
GenBank mRNA | AK009000.1 | - | 5 - 6 | 797 | 72 |
BC002100.1 | - | 5 - 6 | 811 | 72 |