Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm20605
- Entrez Gene ID
-
100316903
- Gene Name
- predicted gene 20605
- Gene Aliases
-
Lrch4-sap25
- Chromosome Location
-
Chr.5: 137629123 - 137642896 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033148.1 |
|
18 - 19 |
3800 |
85 |
GenBank mRNA |
AK164339.1 |
- |
1 - 2 |
1007 |
85 |
- Gene Symbol
- Lrch4
- Entrez Gene ID
-
231798
- Gene Name
- leucine-rich repeats and calponin homology (CH) domain containing 4
- Gene Aliases
-
2810008P14Rik, 2900069C24Rik, AI558103, LRN, LRRN4, SAP25, mFLJ00248
- Chromosome Location
-
Chr.5: 137629123 - 137641099 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK131167.1 |
- |
- |
3959 |
85 |
- Gene Symbol
- Sap25
- Entrez Gene ID
-
751865
- Gene Name
- sin3 associated polypeptide
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 137641473 - 137642902 on Build GRCm38
View Full Product Details