The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027891.4 | NP_082167.2 | 14 - 15 | 1930 | 74 |
GenBank mRNA | AA620153.1 | - | 1 - 2 | 71 | 74 |
AK004717.1 | - | 12 - 13 | 2892 | 74 | |
AK084948.1 | - | 14 - 15 | 1930 | 74 | |
AK085327.1 | - | 3 - 4 | 1961 | 74 | |
AK153976.1 | - | 14 - 15 | 1901 | 74 | |
AK154352.1 | - | 14 - 15 | 1873 | 74 | |
AK167020.1 | - | 14 - 15 | 1879 | 74 | |
AK167868.1 | - | 2 - 3 | 279 | 74 | |
AK172708.1 | - | 14 - 15 | 1899 | 74 | |
AK208344.1 | - | 1 - 2 | 113 | 74 |