Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb5
Entrez Gene ID
81007
Gene Name
defensin beta 5
Gene Aliases
BD-5
Chromosome Location
Chr.8: 19247592 - 19250828 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030734.2 NP_109659.2 1 - 2 60 81

View Full Product Details