Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam32
Entrez Gene ID
353188
Gene Name
a disintegrin and metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 24836143 - 24948917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293693.1 NP_001280622.1 1 - 2 85 155
NM_153397.2 NP_700446.2 1 - 2 85 155
GenBank mRNA AF513715.1 - 1 - 2 68 155
AK005759.1 - 1 - 2 76 155
AK131749.1 - 1 - 2 85 155
BC060983.1 - 1 - 2 82 155

View Full Product Details