Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam32
Entrez Gene ID
353188
Gene Name
a disintegrin and metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 24836143 - 24948917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293693.1 NP_001280622.1 6 - 7 539 134
NM_153397.2 NP_700446.2 6 - 7 539 134
GenBank mRNA AF513715.1 - 6 - 7 522 134
AK005759.1 - 6 - 7 530 134
BC060983.1 - 6 - 7 536 134

View Full Product Details