The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033225.2 | NP_001028397.2 | 1 - 2 | 836 | 74 |
GenBank mRNA | AK159693.1 | - | 1 - 2 | 559 | 74 |