The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022656.2 | NP_073147.2 | 15 - 16 | 1871 | 56 |
GenBank mRNA | AF315344.1 | - | 11 - 12 | 1306 | 56 |
AK122412.1 | - | 13 - 14 | 1387 | 56 | |
BC089329.1 | - | 10 - 11 | 1090 | 56 | |
BC108364.1 | - | 10 - 11 | 1066 | 56 | |
BC144980.1 | - | 11 - 12 | 1200 | 56 |