The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291432.1 | NP_001278361.1 | 5 - 6 | 1642 | 76 |
NM_001291433.1 | NP_001278362.1 | 5 - 6 | 1694 | 76 | |
NM_011805.3 | NP_035935.2 | 5 - 6 | 1694 | 76 | |
NM_175551.4 | NP_780760.2 | 5 - 6 | 1694 | 76 | |
NM_177852.4 | NP_808520.2 | 5 - 6 | 1642 | 76 | |
NR_111965.1 | 4 - 5 | 1552 | 76 | ||
GenBank mRNA | AJ238332.1 | - | 5 - 6 | 1596 | 76 |
AK042474.1 | - | 5 - 6 | 1663 | 76 | |
AK129117.1 | - | 5 - 6 | 1642 | 76 | |
AY425951.1 | - | 5 - 6 | 1596 | 76 | |
AY425952.1 | - | 5 - 6 | 1596 | 76 | |
BC060249.1 | - | 5 - 6 | 1643 | 76 | |
BC096662.1 | - | 5 - 6 | 1634 | 76 | |
BC138712.1 | - | 4 - 5 | 1471 | 76 | |
BC138713.1 | - | 4 - 5 | 1471 | 76 | |
BC145401.1 | - | 4 - 5 | 1472 | 76 |